Would you like to log in again?
HarvestLine System Liners, Beckman Coulter
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
369256 C15220 C26740 C15121 C26742 C15219 358299 369256 369264 C15121 369264 C15124 361690 C15124 C19681 C15218 361690 C15218 C15220 C26740 C19681 C15221 C15217 C15217 356855 C15221 C26742 C15216 358299 C15219 356855 C15216
BK356855PK 182.99 USD InStock
BK356855 CA76292-818 CABK358299 BK358299 76292-130 76292-132 CA76292-140 CA76292-080 CA76292-082 CABK356855 76294-184 76294-186 CA76292-820 76292-134 CABK369264 CA76292-142 76292-136 BK369264 76292-064 CA76292-072 76292-066 CA76292-074 76292-060 CA76292-070 76292-062 CABK369256 BK361690 76292-128 CA76292-076 CABK361690 CA76292-078 BK369256
HarvestLine System Liners, Beckman Coulter
Bags Media Bags
Faster, safer, more efficient harvesting and now it's all in the bag. Minimize non-value-added tasks e.g. bottle cleaning, autoclaving and scraping. Reduce exposure to potentially harmful biologics.
  • Prevent contamination
  • Reduce repetitive motions
  • Prevent liquid spillage
  • Reduce needed storage space
  • Achieve maximum ample yield
  • Expedite freeze/thaw lysing
Safe, efficient workflows are as critical to study as quality samples and to harvest high-quality samples in fewer steps from prep to cleanup.
Optional sterile bottle liners, optimized speed, safety and pelleting efficiency are in the bag.
Different options of HarvestLine System Liners available, also called HarvestLine bottle liners, to address the research needs: 500, 750 and 1000 ml, and sterile and non sterile.
Ordering information: 500, 750 and 1000 ml Beckman Coulter bottles can be purchased separately and accomodate the corresponding HavestLine system/bottle liners.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty