Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Digi-Sense® Temp300 Dual-Input Data Logging Thermocouple Thermometer, Cole-Parmer
Supplier: Cole Parmer
91428-04
76292-168EA 887.87 USD InStock
76292-168
Digi-Sense® Temp300 Dual-Input Data Logging Thermocouple Thermometer, Cole-Parmer
Data Loggers
Digi-Sense® Temp300 dual-input data logging thermocouple thermometer log up to 2000 sets of readings in real time, with time and date stamp. Large easy-to-read screen with back light
  • Three way hands-free operation available
  • Menu-driven set up to easily customize your meter
  • Transfer data via USB
  • Rugged armor with built-in stand—withstands the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Low battery indicator
  • Three years warraanty
Thermocouple measurement type.
Certifications: CE Certified.
Ordering information: Probes are sold separately.
Delivery information: Includes protective rubber armor/boot with stand and three AA batteries.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty