Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Digi-Sense® Temp340 Single-Input Data Logging Thermistor Thermometer, Cole-Parmer
Supplier: Cole Parmer
91428-06
76292-172EA 873.45 USD InStock
76292-172
Digi-Sense® Temp340 Single-Input Data Logging Thermistor Thermometer, Cole-Parmer
Data Loggers
This thermometer offers versatility and accuracy in a compact design. Log up to 2000 sets of readings in real time, with time and date stamp.
  • Three way hands-free operation available
  • Menu-driven set up to easily customize your meter
  • Transfer data via USB
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Low battery indicator
  • Three years warranty
Thermistor measurement type. Ergonomically designed for both large and small hands. Ideal for use in food processing plants, research and science labs, building maintenance areas.
Certifications: CE certified.
Ordering information: Probes are sold separately.
Delivery information: Includes protective rubber armor/boot with stand and three AA batteries.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty