Would you like to log in again?
LOT SPECIFIC CALIBRATE SOFTWARE F/DR3900
Supplier: Hach
Lot Specific Calibrate Software F/Dr3900^ Truecal* Lot Specific Calibration Software, for DR3900/DR6000, reduces variations in results caused by variances in chemistry raw Materials, bar code contains the calibration curve data specific to each batch of c
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LOT SPECIFIC CALIBRATE SOFTWARE F/DR3900 76292-190 LPZ440.99.00001 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty