Would you like to log in again?
CAP SCREW WITH BLUE GASKET PP CASE OF960
Supplier: Greiner Bio-One
Cap Screw With Blue Gasket Pp Case Of960^ Screw Cap, Material: Polypropylene, medical grade and USP class VI certified, with Blue gasket, Gasket Material: Silicone, Free of detectable DNase/Rnase, human DNA and pyrogens, Contents non-cytotoxic, 96 pieces/
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP SCREW WITH BLUE GASKET PP CASE OF960 76292-200 385270 Case of 960 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty