Would you like to log in again?
RACK W/LID PC BLACK F/300UL TUBES CASE10
Supplier: Greiner Bio-One
Rack W/Lid Pc Black F/300Ul Tubes Case10^ Rack, with Rack Lid, Material: Polycarbonate, (Rack/lid), Colour: black(rack), transparent(lid), non-sterile, 1D/2D code, for 300 ul Cryo.s* Biobanking Tubes, stackable, with a footprint that corresponds of the Am
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RACK W/LID PC BLACK F/300UL TUBES CASE10 76292-222 976501 Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty