Would you like to log in again?
RACK W/96 CAP TUBE PC BLUE 600UL CS/960
Supplier: Greiner Bio-One
Rack W/96 Cap Tube Pc Blue 600Ul Cs/960^ Rack, with 96 Cryo.s* Biobanking 96 capped Tubes, with Screw cap, Sterile, Material: Polypropylene(tube/cap), Polycarbonate(rack/lid), Gasket: Silicone, medical grade, Color: blue(cap), black(rack), 2 racks/bag, 1
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RACK W/96 CAP TUBE PC BLUE 600UL CS/960 76292-242 977564 Case of 960 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty