Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Open End Serological Pipettes

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
GSP310001 GSP311001 GSP311005 GSP311010 GSP310005 GSP311002 GSP310002 GSP310010
76292-574CS 119.26 USD InStock
76292-574 76292-288 76292-576 76292-294 76292-296 76292-286 76292-290 76292-292
VWR® Open End Serological Pipettes
Pipettes Serological Pipettes
Wide, open end design is ideal for viscous and high particulate solutions, as well as slurries such as Sephadex.

  • Temperature range: –20 to 50 °C (–4 to 122 °F)
  • Pipettes are latex-free
  • Sterilized by irradiation
  • DNase-/RNase-free, non-pyrogenic, and non-cytotoxic
  • Graduated in 1/10 ml
  • Pipettes are not autoclavable
  • Dispenser box for easy opening

Shelf life: 3 years after month of production.

Graduations are calibrated for accurate dispensing (within ±2%). Negative graduations up to –0.3 ml (total volume: 1.3 ml).

All pipettes are printed with a batch number for traceability. Meets BSE/TSE requirements.

Certifications: Manufactured in a class 100000 cleanroom environment. Manufactured under EN ISO13485 and ISO9001 quality management systems.

Packaging: Pipettes are available individually packed (in peel-to-open wraps with lot number for quality traceability), or bulk packed in easy-to-open bags.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty