Would you like to log in again?
Gas Cylinder Forklift Pallets with Firewall, End Loaded, Justrite®
Supplier: Justrite
35222
76292-346EA 4148.19 USD InStock
76292-346
Gas Cylinder Forklift Pallets with Firewall, End Loaded, Justrite®
Drum Storage
These Gas Cylinder Forklift Pallets with Firewall provide safety when moving and storing of gas cylinders.
  • Steel encased Marinite® I creates a 1 hour fire barrier between the fuel and oxidizer
  • Fully enclosed 197 mm (7.75") wide forklift channels with two-way access
  • Textured finish on the pallet provides a non-skid surface
  • Secure transportation of multiple gas cylinders with a forklift using these end-load oriented pallets
  • Made of steel encased Marinite.
Secure a group of gas cylinders using the two included 4,536 kg (10000 lbs.) DOT compliant straps that attach to the frame at either 305 mm, 610 mm or 915 mm (12", 24" or 36") from the deck.
Fully welded construction from 11 gauge (3 mm) hot-rolled steel and structural tube steel, powder-coated to ensure longevity and chemical resistance.
Certifications: Forklift pallets are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty