Would you like to log in again?
Gas Cylinder Barricade Rack Pallet with Ramp, Justrite®
Supplier: Justrite
35224 35226
76292-348EA 2721.94 USD InStock
76292-348 76292-350
Gas Cylinder Barricade Rack Pallet with Ramp, Justrite®
Drum Storage
These Gas Cylinder Barricade Rack Pallets with Ramp provide safety when moving and storing of gas cylinders.
  • Secure cylinders up to 279 mm (11") in diameter
  • Fully enclosed 197 mm (7.75") wide forklift channels with two-way access
  • Secure transportation of multiple gas cylinders with a forklift using these end-load oriented barricade rack pallets
Barricade racks surround cylinders with structural steel tubing, continuously seam welded at all junction points and mounted to the pallet.
They are sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability. Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries.
Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit.
Certifications: Rack pallets are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty