Would you like to log in again?
Pneumatic Wheels and Axle Set for Single Gas Cylinder Hand Trucks, 10.5", Justrite®
Supplier: Justrite
35374
76292-452EA 148.46 USD InStock
76292-452
Pneumatic Wheels and Axle Set for Single Gas Cylinder Hand Trucks, 10.5", Justrite®
Carts
Pneumatic Wheels and Axle Set for Single Gas Cylinder Hand Trucks provides safety when moving and storing of gas cylinders.
  • Wheel and axle replacement set for single cylinder Justrite® hand trucks
  • Wheels are 10.5×4"
  • Axel size is 17.75×0.625"
Delivery information: Includes two 10.5" pneumatic wheels with welded steel hubs and dual ball bearings, and plated axle with clips and washers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty