Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Double Cylinder Hand Truck with Hoist Ring and Locking Post, Justrite®

Supplier: Justrite

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

35024 35034 35026 35018 35022
76292-496EA 1632.61 USD InStock
76292-496 76292-486 76292-488 76292-484 76292-480
Double Cylinder Hand Truck with Hoist Ring and Locking Post, Justrite®
Carts
This Double Cylinder Hand Truck with Hoist Ring and Locking Post provides safety when moving and storing of gas cylinders.

  • Durable
  • Provide optimum safety, strength, and longevity
  • Developed for the US Navy for use on submarines
  • Locking posts and collar plates ensure total cylinder immobilization
  • Corrosion-resistant powder-coat finish throughout for smooth handling
  • Floor plate made of thick 11 gauge (3 mm) steel with safety fins prevents warping or breaking
  • Accessorize hand trucks with a tool tray or lockable tool box for securing torches and other equipment

Move gas cylinders easily around a construction site, into locations with poor accessibility, or from floor to floor with these hoist-equipped hand trucks.

Extensive structural framing with 10 mm (0.375") thick hoist plates make these units safe when lifted by hoist rigs. Triple grip handle design helps user control the high center of gravity by expanding available grip area for improved leverage and swing radius.

The zero-lift height loading allows floor plate to lay flat, thereby eliminating the need to lift cylinder during loading and unloading.

Caster carriage model features heavy steel bars and nylon washers for smooth folding action and easy storage in tight spaces.

Two heavy-duty 38×1372 mm (1.5×54") polypropylene support straps, lab tested to 1138 lbs. (516 kg), feature steel cinch buckles for secure attachment of cylinder. Straps eliminate any cylinder rolling potential seen in chain-secured units.

Certifications: Hand Trucks are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty