Would you like to log in again?
Lift-and-Load Single Cylinder Hand Truck, Justrite®
Supplier: Justrite
35054
76292-516EA 1602.78 USD InStock
76292-516
Lift-and-Load Single Cylinder Hand Truck, Justrite®
Carts
Lift-and-Load Single Cylinder Hand Truck provides safety when moving and storing of gas cylinders.
  • Built-in clamping system with welded loop chain secures the cylinder for lifting and transport
  • Accepts cylinder sizes from 102 to 305 mm (4 to 12") in diameter
  • Fully welded construction, protected with durable, corrosion-resistant, red powder-coat finish
Designed to lift, transport, and install gas cylinders on welding carts, platforms, pallets, and in cabinets or other elevated surfaces.
The manual lift system has a jack with an 279 mm (11") extension geared to make lifting easy, with approximately 10 turns of the crank per vertical inch.
Certifications: Hand Trucks are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Delivery information: Includes pneumatic 267 mm (10.5") main wheels and wide-set, dual (3") 5 mm rear swivel casters for increased stability and maneuverability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty