Would you like to log in again?
VENTILATED BALANCE ENCLOSURE 36X28X41IN
Supplier: PLAS-LABS
Ventilated Balance Enclosure 36X28X41In^ Ventilated Balance Enclosure, Dimensions: 36x28x41in, with top mounted HEPA filter and blower, air flow alarm system, phenolic base, electrical cord outlet port, 6in exhaust duct, and optional bag-in / bag-out port
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VENTILATED BALANCE ENCLOSURE 36X28X41IN 76292-828 900-VBE/36 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty