Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Grade 1V Qualitative Filter Paper, Fluted

Supplier: Whatman products (Cytiva)
Whatman™
10380405 1201-185 1201-270 1201-150 1201-320 10380406 10380404 1201-125 1201-125 1201-185 10380405 1201-270 10380406 1201-150 10380404 1201-240 1201-240 1201-320
10828-396PK 135.85 USD InStock
10828-396 CA10828-394 10828-394 CA10828-396 CA10827-846 10827-848 76292-874 CA10827-988 CA76292-872 CA10827-848 CA76292-874 10827-844 76292-870 10827-846 CA10827-844 76292-872 10827-988 CA76292-870
Whatman™ Grade 1V Qualitative Filter Paper, Fluted
Filters Filter Paper Qualitative Filter Paper
Whatman Grade 1V lab filter paper from Cytiva business is a widely used pre-folded filter for many general filtration applications.

  • Nominal particle retention rating of 11 µm supports general filtration
  • Intermediate filtration speed (Herzberg 150 s)
  • Pre-folded filter for ease-of-use and time savings with conical filter funnels; avoids the need to quadrant-fold circles

Cytiva offers a full line of qualitative papers. As the pleated and pre-folded version of Whatman filter paper 1, Grade 1V is well suited for qualitative analysis in routine applications including: Separations of lead sulfate, hot calcium oxalate, and calcium carbonate; Agriculture, for soil analysis; Food quality testing to separate solid foodstuffs from associated liquid; Education, to teach simple qualitative analysis.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty