Would you like to log in again?
2'3'-cGAMP ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical
501700-96
76292-890EA 622.16 USD InStock
76292-890
2'3'-cGAMP ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of 2’3’-cGAMP in cell lysates.
  • Measure 2'3'-cGAMP in cell lysates
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Lower limit of detection (LLOD) is 9.6 pg/ml (0.01 pmol/ml)
  • Run overnight or incubate for just 2 hours without compromising sensitivity
  • Monitor the kinetics of 2'3'-cGAMP formation and hydrolysis in a biological setting
  • Identify compounds that modulate cGAS activity
Cayman’s 2’3’-cGAMP ELISA Kit is a competitive assay that can be used for quantification of 2’3’-cGAMP in cell lysates. This assay has a range from 100 ng/ml to 6.1 pg/ml with a midpoint of approximately 900 pg/ml (50% B/B0) and a sensitivity of approximately 85 pg/ml (80% B/B0). Powered by BIOLOG Life Science Institute.
Delivery information: Kit contains: Antiserum, tracer, standard, immunoassay buffer C concentrate, wash buffer concentrate, Polysorbate 20, coated plate, substrate solution, stop solution, tracer dye, antiserum dye, and cover sheet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty