Would you like to log in again?
Human Recombinant cGAS (from E. coli)
Supplier: Cayman Chemical
25001-100
76292-894EA 509.15 USD InStock
76292-894
Human Recombinant cGAS (from E. coli)
Enzymes
Cyclic GMP-AMP (cGAMP) synthase (cGAS) (161-522) is a truncated form of cGAS (Item No. 22810) that contains the nucleotidyltransferase and Mab21 domains. cGAS is a nucleotidyltransferase located in the cytosol that acts as a cytosolic DNA sensor to detect foreign DNA from microbial pathogens as part of the innate immune response. Upon binding to cytosolic DNA, cGAS produces the cyclic dinucleotide second messenger cGAMP, which activates stimulator of interferon genes (STING), leading to activation of the type I interferon (IFN) pathway. <em>In vitro</em>, fibroblasts, macrophages, and dendritic cells isolated from cGAS knockout (cGAS<sup>-/-</sup>) mice do not produce type I IFNs following DNA transfection or DNA virus infection. Similarly, cells containing a frame-shift mutation in the cGAS locus fail to mount an immune response to HIV and other retroviruses. <em>In vivo</em>, cGAS<sup>-/-</sup> mice infected with herpes simplex virus 1 (HSV-1) have lower levels of IFN-α and IFN-β, shorter survival times, and higher post-mortem levels of HSV-1 in the brain.
  • UniProt: Q8N884
Used for: Innate Immunity, Infectious Disease, Intracellular Signaling
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty