Would you like to log in again?
Human Recombinant CdnP (from M. Tuberculosis strain ATCC 25618/H37Rv)
Supplier: Cayman Chemical
22809-100
76292-930EA 472.45 USD InStock
76292-930
Human Recombinant CdnP (from M. Tuberculosis strain ATCC 25618/H37Rv)
Enzymes
Cyclic di-nucleotide phosphodiesterase (CdnP, also known as Rv2837c) is a soluble, stand-alone phosphodiesterase in regulating cyclic dinucleotide signaling during intracellular infections of <em>M. tuberculosis</em>{36095}. Studies have found that CdnP is capable of hydrolyzing cyclic di-adenosine monophosphate (c-di-AMP; Item No. 17753) into two AMPs as a strategy to avoid activation of the innate immune response of the host. <em>M. tuberculosis</em> infection leads to cytosolic release of c-di-AMP, which is recognized by stimulator of interferon genes (STING) and subsequently triggers type I interferon response. Other studies have shown that deletion or inhibition of CdnP attenuates the <em>M. tuberculosis</em> virulence both <em>in vitro</em> and <em>in vivo</em>.
Used for: Antivirals, Autoimmunity, Infectious Disease, Innate Immunity, Tuberculosis Research
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty