Would you like to log in again?
Human Recombinant STING H232 variant (from E. coli)
Supplier: Cayman Chemical
22815-100
76293-152EA 741.3 USD InStock
76293-152
Human Recombinant STING H232 variant (from E. coli)
Enzymes
  • UniProt: Q86WV6
Stimulator of interferon genes (STING) is a component of the innate immune response that binds to cyclic dinucleotides, which are bacterial second messengers, leading to activation of NF-κB and transcription of immunomodulatory genes, including type I interferon (IFN). The H232 variant of STING is found at a 13.7% frequency in the 1000 Genome Project. The SNP variant R232 (Item No. 22816) is the most common variant in the human population, found at a frequency of 57.9%. Various mutations in STING either reduce or increase its activity. Gain-of-function mutations in STING, including R284M (Item No. 23594) and V155M, lead to constitutive activation and enhancement of the type I IFN response. The V155M mutation is associated with a systemic inflammatory condition, including pulmonary fibrosis and autoimmune factors. Mutations that reduce STING activity include K224R (Item No. 23593), which reduces ubiquitination of STING thereby disrupting its localization within the cell, and the double mutation G230A, R293Q (Item No. 23592), which reduces the IFN response. A T596A mutation present in the mouse strain Goldenticket leads to a complete loss of STING protein and lack of a type I IFN response to infection by Listeria.
Used for: Infectious Disease, Autoimmunity, Signal Transduction
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty