Would you like to log in again?
NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina, New England Biolabs
Supplier: New England Biolabs
NEBNext®
E6420S E6420L E6420S E6420L
76293-216EA 1311 USD InStock
76293-216 CA76293-214 76293-214 CA76293-216
NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina uses a template switching method to generate full length cDNAs directly from single cells or 2 pg to 200 ng RNA, followed by conversion to sequence-ready libraries using the Ultra II FS workflow.
  • Experience unmatched detection of low abundance transcripts
  • Rely on consistent transcript detection for a wide range of input amounts and sample types
  • Observe uniform transcript coverage, regardless of input amount or sample type
  • Use with a variety of RNA inputs, including cultured or primary cells, or total RNA
This unique workflow enables generation of the highest yields from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance.
Ordering information: Please note that adaptors and primers are not included in the kit and are available separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty