Would you like to log in again?
Stamped Steel Heavy Support Stands with Rods, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SSB4X6-H-VWR SSB6X9-H-VWR SSB5X8-H-VWR SSB6X1-H-VWR
76293-244EA 23.19 USD InStock
76293-244 76293-242 76293-248 76293-246
Stamped Steel Heavy Support Stands with Rods, United Scientific Supplies
Support Stands
Heavy stamped steel bases with black enamel finish feature an added steel plate on the back, which makes them about 50% heavier than regular stamped steel bases.
  • Zinc plated rod
The steel plate also facilitates a sturdier threading of the rod into the base.
Ordering information: Bases and Rods are also available individually.
Delivery information: Includes a removable, zinc-plated steel rod.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty