Would you like to log in again?
Stamped Steel Two-Hole Support Stands with Rods, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SSB6X9-H2-VWR SSB4X6-H2-VWR SSB5X8-H2-VWR SSB6X1-H2-VWR
76293-256EA 32.29 USD InStock
76293-256 76293-254 76293-252 76293-250
Stamped Steel Two-Hole Support Stands with Rods, United Scientific Supplies
Support Stands
Tip-free stability for experiments.
  • Features two tapped holes
  • zinc plated rod
Stamped steel bases with black enamel finish features 2 tapped holes. Hole at center of the shorter side is for standard ring stand use. Hole at center of the longer side is for applications such as titrations, specific gravity determinations and more.
Ordering information: Bases and Rods are also available individually.
Delivery information: Includes a removable, zinc-plated steel rod.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty