Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

A-Base and Rod, Cast Iron, United Scientific Supplies

Supplier: United Scientific Supplies
SBA724-VWR
76293-262EA 38.48 USD InStock
76293-262
A-Base and Rod, Cast Iron, United Scientific Supplies
Support Stands
Compact design to hold equipment securely.

  • Shipping weight: 4 lbs.
  • Dimensions: 24×9×2"

The A-shaped base is made of rugged, cast iron and is designed for extra stability. It has 8" long legs with a black enamel finish. The 5" to 7.5" opening between the legs is designed for positioning items near the support rod.

Delivery information: Includes a removable, zinc-plated steel rod that is 24" long and 0.5" in diameter.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty