Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Extension Support Rings, United Scientific Supplies
Supplier: United Scientific Supplies
SRSE02-VWR SRSE06-VWR SRSE04-VWR SRSE03-VWR SRSE05-VWR
76293-662EA 4.34 USD InStock
76293-662 76293-342 76293-336 76293-338 76293-340
Extension Support Rings, United Scientific Supplies
Support Rings
Extension support for holding funnels.
  • Steel rod, zinc plated
  • 12" long
Steel rod, zinc-plated extension support rings. Rod diameter 5/16", overall length 12" for all sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty