Would you like to log in again?
Solid Rubber Stoppers, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RST10-S-VWR RST0-S-VWR RST00-S-VWR RST7A-S-VWR RST11A-S-VWR RST8-S-VWR RST11-S-VWR RST9A-S-VWR RST10A-S-VWR RST4-S-VWR RST9-S-VWR RST7-S-VWR RST2-S-VWR RST6-S-VWR RST5A-S-VWR RST14-S-VWR RST1-S-VWR RST5-S-VWR RST3-S-VWR RST12-S-VWR RST15-S-VWR RST16-S-VWR RST6A-S-VWR RST13-S-VWR RST000-S-VWR RST8A-S-VWR
76293-476EA 9.81 USD InStock
76293-476 76293-498 76293-474 76293-496 76293-352 76293-512 76293-478 76293-510 76293-490 76293-472 76293-494 76293-350 76293-492 76293-486 76293-484 76293-502 76293-348 76293-500 76293-488 76293-664 76293-506 76293-504 76293-508 76293-280 76293-482 76293-480
Solid Rubber Stoppers, United Scientific Supplies
Stoppers Rubber Stoppers
Solid, black stoppers contain a high percentage of pure rubber and weigh less than neoprene.
  • Resilient and pliable for a long time
  • Low in free sulfur
  • Can withstand adverse laboratory conditions without hardening
These black stoppers are made of solid natural rubber that stays pliable over a long period of time. Ideal for use with non-aggressive or concentrated solutions. Low sulfur content. Stoppers will withstand temperatures from –25 to 70 °C (–13 to 158 °F). Each stopper has its size marked in raised figures.
Packaging: Sold as one-pound bags.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty