Would you like to log in again?
Burette Clamp, Stainless Steel, United Scientific Supplies
Supplier: United Scientific Supplies
CLBUR1N-VWR CLBUR2N-VWR
76293-398EA 12.91 USD InStock
76293-398 76293-396
Burette Clamp, Stainless Steel, United Scientific Supplies
Clamps General Purpose Clamps
These clamps are constructed from steel and feature plastic covered spring jaws.
  • Available in single and double versions
These clamps accept burettes up to 35 mm O.D., and can be used with support rods up to 1/2" O.D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty