Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brass Clamp Holder, United Scientific Supplies

Supplier: United Scientific Supplies
RCLBH2-VWR
76293-414EA 12.3 USD InStock
76293-414
Brass Clamp Holder, United Scientific Supplies
Clamps General Purpose Clamps
Easily connect items to a support rod.

  • Thumb screws 180° apart
  • Hold support rods up to 3/4" OD at a 90° angle

This all brass clamp holder allows you to connect support rods together up to 19 mm in diameter.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty