Would you like to log in again?
Vigreux Distilling Columns, 24/40 Joint, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CDV500-VWR CDV300-VWR CDV400-VWR CDV200-VWR
76293-464EA 94.58 USD InStock
76293-464 76293-462 76293-458 76293-460
Vigreux Distilling Columns, 24/40 Joint, United Scientific Supplies
Distillation Components Distillation Columns
Vigreux distilling columns for cooling and condensing.
  • Features downward pointing indentations to improve cooling
  • 24/40 sockets
A Vigreux column is a type of air condenser where the simple tube has been modified to include an abundance of downward-pointing indentations. This dramatically increases the surface area per unit length of the condenser.
Delivery information: Includes a 24/40 connector.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty