Would you like to log in again?
Porcelain Crucibles with Porous Bottom, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
JCP25050-VWR JCP25012-VWR JCP25150-VWR
76293-546EA 303.87 USD InStock
76293-546 76293-544 76293-548
Porcelain Crucibles with Porous Bottom, United Scientific Supplies
Crucibles
Porous bottom crucibles have a molded-in porous disc with stable porosity and consistent flow rate.
  • Porous bottom
  • Glazed
Available with very fine (1.2 µm), fine (5 µm), and medium (15 µm) pore diameters. Glazed inside and out, except for the outside bottom and rim. Autoclavable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty