Would you like to log in again?
Lattice Lab System Set #4, United Scientific Supplies
Supplier: United Scientific Supplies
LLS004-VWR
76293-588EA 1.2 USD InStock
76293-588
Lattice Lab System Set #4, United Scientific Supplies
Supports Flask Supports
This Lattice Lab System set features aluminum rods up to 72" long.
  • Sturdy aluminum construction
  • Includes mounting hardware to tables
Assembly is easy and requires only a screwdriver. Support rods are made of hard aluminum, and foot plates and hook connectors are made of cast alloy.
Delivery information: Set #4 includes: (8) Foot Plates, 2.3125" diameter; (8) Set Screws for Foot Plate; (24) Anchoring Screws for Foot Plate; (49) Hook Connectors with Thumbscrew; (6) Support Rods, 2" L, 0.5" diameter; (6) Support Rod, 12" L, 0.5" diameter; (2) Support Rod, 24" L, 0.5" diameter; (7) Support Rod, 48" L, 0.5" diameter; (5) Support Rod, 72" L, 0.5" diameter.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty