Would you like to log in again?
Laboratory Jack, Anodized Aluminum and Stainless Steel, United Scientific Supplies
Supplier: United Scientific Supplies
LBJK01-VWR
76293-612EA 103 USD InStock
76293-612
Laboratory Jack, Anodized Aluminum and Stainless Steel, United Scientific Supplies
Laboratory Jacks
Laboratory support jack is constructed of chemically resistant anodized aluminum and stainless steel.
  • Chemically resistant
  • Stainless steel and aluminum construction
The scissor jack design enables height adjustment with a simple turn of the knob. Sturdy design can support loads up to 20 kg. Platform height can be vertically adjusted from closed position of 2.75" up to 10.5".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty