Would you like to log in again?
Support Stand and Ring Sets, United Scientific Supplies
Supplier: United Scientific Supplies
SET694-VWR SET583-VWR SET462-VWR SET615-VWR
76293-630EA 39.28 USD InStock
76293-630 76293-634 76293-632 76293-636
Support Stand and Ring Sets, United Scientific Supplies
Support Stands
Each set contains a stamped steel base and a zinc-plated steel rod, along with 2, 3, 4, or 5 cast iron rings with clamps.
  • Sets comes with 2, 3, 4 or 5 rings of varied diameters
  • Base is stamped steel and the plate underneath is made of steel
  • Base weighs 2 lbs.
The support stand will support up to 10 lbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty