Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cast Iron Support Rings, United Scientific Supplies
Supplier: United Scientific Supplies
SRCI06-VWR SRCI05-VWR SRCI03-VWR SRCI07-VWR SRCI04-VWR SRCI02-VWR
76293-640EA 9.7 USD InStock
76293-640 76293-644 76293-642 76293-638 76293-648 76293-646
Cast Iron Support Rings, United Scientific Supplies
Support Rings
Support rings for funnels of all sizes.
  • Cast iron, zinc plated
  • Boss head clamp
Cast iron, zinc-plated support ring with boss head clamp and thumbscrew. Distance from support to center of ring is 4-1/2" for all sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty