Would you like to log in again?
Rubber Stoppers, 1 Hole, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RST16-H-VWR RST7A-H-VWR RST00-H-VWR RST9-H-VWR RST4-H-VWR RST10A-H-VWR RST6A-H RST7-H-VWR RST8-H-VWR RST3-H-VWR RST5A-H-VWR RST6A-H-VWR RST0-H-VWR RST1-H-VWR RST14-H-VWR RST12-H-VWR RST11A-H-VWR RST5-H-VWR RST11-H-VWR RST8A-H-VWR RST9A-H-VWR RST000-H-VWR RST6-H-VWR RST15-H-VWR RST10-H-VWR RST2-H-VWR RST13-H-VWR
76293-696EA 10.16 USD InStock
76293-696 76293-674 76293-672 76293-694 76293-678 76293-710 76293-676 76293-698 76293-714 76293-712 76293-716 76293-692 76293-670 76293-690 76293-684 76293-682 76293-700 76293-688 76293-666 76293-686 76293-704 76293-702 76293-668 76293-708 76293-706 470005-776 76293-680
Rubber Stoppers, 1 Hole, United Scientific Supplies
Stoppers Rubber Stoppers
Solid, black stoppers contain a high percentage of pure rubber and weigh less than neoprene.
  • Resilient and pliable for a long time
  • Low in free sulfur
  • Can withstand adverse laboratory conditions without hardening
These black stoppers are made of solid natural rubber that stays pliable over a long period of time. Ideal for use with non-aggressive or concentrated solutions. Low sulfur content. Stoppers will withstand temperatures from –25 to 70 °C (–13 to 158 °F). Each stopper has its size marked in raised figures.
Packaging: Sold as one-pound bags.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty