Would you like to log in again?
Rubber Stoppers, 2 Hole, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RST2-H2-VWR RST6-H2-VWR RST9A-H2-VWR RST4-H2-VWR RST00-H2-VWR RST13-H2-VWR RST5-H2-VWR RST1-H2-VWR RST6A-H2-VWR RST0-H2-VWR RST9-H2-VWR RST10A-H2-VWR RST10-H2-VWR RST11A-H2-VWR RST12-H2-VWR RST3-H2-VWR RST14-H2-VWR RST15-H2-VWR RST5A-H2-VWR RST7-H2-VWR RST7A-H2-VWR RST16-H2-VWR RST11-H2-VWR RST8A-H2-VWR RST8-H2-VWR
76293-750EA 9.74 USD InStock
76293-750 76293-732 76293-754 76293-730 76293-752 76293-736 76293-758 76293-734 76293-756 76293-718 76293-738 76293-762 76293-740 76293-760 76293-722 76293-766 76293-744 76293-720 76293-764 76293-742 76293-748 76293-726 76293-724 76293-746 76293-728
Rubber Stoppers, 2 Hole, United Scientific Supplies
Stoppers Rubber Stoppers
Solid, black stoppers contain a high percentage of pure rubber and weigh less than neoprene.
  • Resilient and pliable for a long time
  • Low in free sulfur
  • Can withstand adverse laboratory conditions without hardening
These black stoppers are made of solid natural rubber that stays pliable over a long period of time. Ideal for use with non-aggressive or concentrated solutions. Low sulfur content. Stoppers will withstand temperatures from –25 to 70 °C (–13 to 158 °F). Each stopper has its size marked in raised figures.
Packaging: Sold as one-pound bags.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty