Would you like to log in again?
Neoprene Stoppers, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NST10-S-VWR NST5-S-VWR NST9-S-VWR NST2-S-VWR NST3-S-VWR NST00-S-VWR NST12-S-VWR NST6-S-VWR NST13-S-VWR NST6A-S-VWR NST000-S-VWR NST11-S-VWR NST8-S-VWR NST5A-S-VWR NST1-S-VWR NSTPK2-VWR NST0-S-VWR NST10A-S-VWR NST11A-S-VWR NST7-S-VWR NST4-S-VWR
76293-784EA 29.59 USD InStock
76293-784 76293-794 76293-772 76293-782 76293-770 76293-792 76293-788 76293-776 76293-798 76293-786 76293-796 76293-774 76293-802 76293-768 76293-778 76293-800 76293-806 76293-804 76293-808 76293-780 76293-790
Neoprene Stoppers, United Scientific Supplies
Stoppers Rubber Stoppers
Synthetic, neoprene stoppers contain no free sulfur and exhibit good resiliency and shelf-life.
  • Neoprene fabrication
Neoprene stoppers can be used in areas where rubber would be attacked by reagents or solvents. Synthetic green neoprene stoppers have good resiliency and shelf-life. They withstand petroleum products and oils as well as most inorganic acids and bases.
Packaging: Sold as one-pound bags.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty