Would you like to log in again?
Rubber Stopper Assortments, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RSTPK1-VWR RSTPK3-VWR RSTPK4-VWR RSTPK2-VWR
76293-810EA 10.56 USD InStock
76293-810 76293-814 76293-812 76293-816
Rubber Stopper Assortments, United Scientific Supplies
Stoppers Rubber Stoppers
Convenient assortments of various sizes of rubber stoppers are sold as one-pound bags.
  • Pliable
  • Autoclavable
Black stoppers are made of solid natural rubber that stays pliable over a long period of time. Ideal for use with non-aggressive or concentrated solutions. Stoppers feature a low sulfur content, and will withstand temperatures from –25 to 70 °C (–13 to 158 °F).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty