Would you like to log in again?
Cork Stoppers, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CST17-VWR CST14-VWR CST11-VWR CST00-VWR CST16-VWR CST10-VWR CST5-VWR CST13-VWR CST6-VWR CST4-VWR CST19-VWR CST18-VWR CST20-VWR CST8-VWR CST2-VWR CST15-VWR CST9-VWR CST000-VWR CST1-VWR CST7-VWR CST0-VWR CST3-VWR CST12-VWR
76293-850PK 160.56 USD InStock
76293-850 76293-860 76293-854 76293-832 76293-820 76293-842 76293-830 76293-852 76293-862 76293-840 76293-858 76293-836 76293-824 76293-846 76293-856 76293-834 76293-844 76293-822 76293-818 76293-828 76293-838 76293-848 76293-826
Cork Stoppers, United Scientific Supplies
Stoppers Cork Stoppers
Cork stoppers function as a closure for bottles, vials, laboratory vessels and other openings.
  • Regular length taper
  • Assorted sizes
These grade XXX tapered cork stoppers are an excellent choice for general laboratory use. They are effective for both liquid and solid mediums and offer light weight, resilience, and compressibility. Chemically inert and moisture-resistant.
Packaging: Sold as packs of 100.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty