Would you like to log in again?
Sapphire Bottle Top Dispenser, United Scientific Supplies
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BTSR100-VWR BTSR2.5-VWR BTSR10-VWR BTSR60-VWR BTSR30-VWR BTSR5-VWR
76293-872EA 657.84 USD InStock
76293-872 76293-882 76293-880 76293-876 76293-874 76293-878
Sapphire Bottle Top Dispenser, United Scientific Supplies
Bottle Top Dispensers
High-quality laboratory instruments for liquid handling needs.
  • Precise
  • Easy to use
  • Autoclavable
Save time and increase lab productivity using the sapphire bottle top dispenser. This convenient dispenser uses a dual valve technology that allows simple rinse and refills without dismounting the dispenser.
User-friendly and high-performance Sapphire Bottle Top Dispensers operate in four modes: Dilution, Rinsing, Dual Liquid Handling, and Re-circulation. Fully autoclavable at 121 °C, 15psi for 10 to 15 minutes.
Certifications: Calibrated ISO 17025.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty