Would you like to log in again?
VWR® Universal Box Holder
Supplier: VWR
VWR®
VWR-76293-884
76293-884CS 943.15 USD InStock
76293-884
VWR® Universal Box Holder
Glove Dispensers and Glove Box Holders
This universal, rotating, and expanding box holder is the only holder that easily expands from 95 to 155 mm (3.75 to 6") to hold a variety of dispensing boxes, containers, and small electronics.
  • Sturdy ABS plastic box holder for spatulas, gloves, lab wipes, and more
  • Unique design provides maximum usability and versatility
  • Holds iPhones, android phones, and small electronic devices
  • Power cord pass through port built in
  • Rotates in 45° increments while mounted
  • Unclutters the bench
  • Made in the USA
The unique VWR® Box Holder features an indexed mounting system, which allows 180° of rotation in 45° increments without removing the box or the holder from the mounting bracket. The unit mounts on most vertical surfaces, such as sheet rock walls, with the included screws and drywall anchors. It can be mounted on the protective lip of lab shelves with the included double-sided tape.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty