Would you like to log in again?
QuantiChrom™ Free Amino Nitrogen Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DFAN-100
76293-886EA 352.06 USD InStock
76293-886
QuantiChrom™ Free Amino Nitrogen Assay Kit, BioAssay Systems
Assays Cellular Assays
Free Amino Nitrogen determination in foods and beverages (e.g. beer, wort, wine, must, etc.).
  • Fast and sensitive: linear detection range (5 µL sample) - 0.2 to 10 mM for 10 min reaction
  • Convenient and high-throughput: homogeneous "mix-incubate-measure" type assay; can be readily automated to process thousands of samples per day
  • Method: OD575 nm
  • Samples: Food and Beverages
FREE AMINO NITROGEN (FAN) is the main source of nitrogen necessary for yeast growth and proper fermentation. Fermentation of beer and wine is processed by yeast, which synthesize proteins using available amino acids. When making beer and wine, free amino nitrogen is extracted from amino acids during the formation of the wort or must.
BioAssay Systems Free Amino Nitrogen assay measures alpha amino acids, ammonia, and end group amino nitrogens. The ninhydrin based reaction is a superior method for determining only alpha amino acids and ammonia compared to the traditional Kjeldahl, which measures nitrogen from all sources. Only requiring low sample volumes, the stable ninhydrin reagent provides a simple and accurate method for determining Free Amino Nitrogen concentrations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty