Would you like to log in again?
EnzyChrom™ Glucose Assay Kit II, BioAssay Systems
Supplier: BioAssay Systems
EGL2-100
76293-888EA 525.35 USD InStock
76293-888
EnzyChrom™ Glucose Assay Kit II, BioAssay Systems
Assays Cellular Assays
For quantitative determination of glucose and evaluation of drug effects on glucose metabolism.
  • Sensitive and accurate: use as little as 20 μl samples; linear detection range in 96-well plate; 0.1 to 3 mM (1.8 mg/dl to 54 mg/dl) glucose for colorimetric assays
  • Convenient: the procedure involves adding a single working reagent, and reading the optical density at 20 minutes; room temperature assay; no 37 °C heater is needed
  • Simple and high-throughput: the procedure involves addition of a single working reagent and incubation for 20 min at room temperature
  • Method: OD340 nm
  • Samples: Serum, plasma, urine, saliva, milk, culture medium, food, beverages etc
Glucose (C6H12O6) is a key diagnostic parameter for many metabolic disorders. Increased glucose levels have been associated with diabetes mellitus, hyperactivity of thyroid, pituitary and adrenal glands. Decreased levels are found in insulin secreting tumors, myxedema, hypopituitarism and hypoadrenalism.
Simple, direct and high-throughput assays for measuring glucose concentrations find wide applications in research and drug discovery. BioAssay Systems glucose assay kit II uses an enzyme to reduce NAD. The produced NADH, measured at 340 nm, is proportional to the glucose concentration in the sample.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty