Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Uritainer™ 24 Hour Urine Collection Containers, Simport Scientific

Supplier: Simport

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

B360-35 B360-25 B360-25
76299-218CS 127.77 USD InStock
CA10836-464 76299-218 76299-216
Uritainer™ 24 Hour Urine Collection Containers, Simport Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
All models feature a unique snap-valve pour spout that is easily popped open to provide dripless pouring.

  • Unique snap-valve pour spout is easily popped open
  • Dripless pouring
  • Specially designed leakproof screw cap with liner for safer transport
  • Metal-free and resistant to hydrochloric acid
  • Large handle can be gripped comfortably
  • Volume level read in 50 ml increments
  • Can be gamma sterilized

This more conventionally styled urine bottle is available in 2.5 and 3.5 l sizes. It also incorporates some of the great features of the URISAFE® 24 Hour Urine Collection Container. The innovative cap has a leakproof design with a cap liner to ensure safe sample transport. The unique snap-valve pour allows spout to easily pop open and push shut. Also offers a controlled flow rate for better handling of poured volumes with reduced risks of aerosol contamination when pouring.

Pour spout eliminates splashing and exposure to hazardous body fluids. A large central handle can be gripped comfortably to reduce fatigue, and permits pouring off of samples with ease.

For IVD use. Chemically resistant. Metal- and latex-free. Zinc- and fluorescence-free. Container can be subjected to freezing, thawing, and EtO gas sterilization without causing changes in materials or physical appearance.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty