Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Antistatic Weigh Dishes, Simport Scientific
Supplier: Simport
D250-2 D250-3 D250-1 D250-2
11006-614CS 49.81 USD InStock
CA11006-614 11006-614 76299-238 76299-236
Antistatic Weigh Dishes, Simport Scientific
Boats Weighing Boats
Weighing Dishes will resist diluted acids, aqueous solutions, alcohols and bases, ideal for many applications such as weighing, dispensing, or storing.
  • Safe, contaminant-free, biologically inert economical containers
  • For weighing liquid or powdered samples in the laboratory
  • Flat bottom ensures perfect stability on countertops
Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, or mixing trays for small batches. Will withstand temperatures up to 80 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty