Would you like to log in again?
StoreBox™ Storage Boxes, Simport
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
StoreBox™
T514-281G T514-281P T514-281B T514-225Y T514-281B T514-2100B T514-225P T514-2100P T514-2100G T514-581Y T514-225Y T514-225B T514-281P T514-281G T514-581G T514-581B T514-2100G T514-2100Y T514-2100B T514-581Y T514-225B T514-581B T514-581P T514-225P T514-281Y T514-581P T514-281Y T514-225G T514-225G T514-2100P T514-581G
76299-318CS 192.75 USD InStock
76299-318 490010-474 CA60872-870 76299-320 CA60872-864 CA60872-866 CA60872-860 CA60872-862 76299-306 490016-354 76299-304 76299-348 490016-352 CA60872-868 CA60872-848 76299-308 CA60872-854 CA60872-876 76299-352 CA60872-856 CA60872-878 76299-350 CA60872-872 CA60872-850 CA60872-852 CA60872-874 76299-316 76299-314 CA60872-858 76299-312 76299-310
StoreBox™ Storage Boxes, Simport
Boxes Storage Boxes
These economical storage boxes are designed to be used at temperatures between –90 and 80 °C. Different models are available to accommodate either 25, 81, or 100 sample tubes.
  • Cover has numbered squares for easy sample identification
  • Two corners of cover and base are cut to prevent misalignment
  • Writing surface provided to identify base and/or cover
  • Vials readily visible through transparent cover
  • Air vents minimize condensation
  • Four pastel colors available for better color coding
A transparent cover allows the user to see the contents of the box. The cover is keyed to the base to prevent misalignment. Printed with a series of squares (numbered), the surface accepts writing with markers, facilitating inventory control.
A unique color coding system uses colored plastic grids to separate the cover from the base on the 25- and 81- place boxes. Those made to accept 100 tubes have a colored base instead of a grid. Vial removal is facilitated by an innovative vial picker supplied with each storage box. Not autoclavable.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty