Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
UVP iBox® Studio touch, in vivo Imaging Systems, Analytik Jena US
Supplier: Analytik Jena
849-97-0933-02 849-97-0932-02 849-97-0933-03 849-97-0933-04 849-97-0932-01 849-97-0932-03 849-97-0933-01 849-97-0932-04
76299-386EA 51980.6 USD InStock
76299-386 76299-396 76299-384 76299-406 76299-404 76299-402 76299-400 76299-398
UVP iBox® Studio touch, in vivo Imaging Systems, Analytik Jena US
Photodocumentation Systems
The UVP iBox Studio is sophistication and ease of use in one system for high resolution small animal fluorescence imaging. From in depth in-vivo studies to quick screening, this powerful imaging system can perform any choice of in-vivo application.
  • Optimize your application by choosing between two high performance cooled camera options
  • Multiple illumination sources – RGBW LEDs and NIR lasers, making the system extremely versatile
  • Included warming plate for up to three mice on an ergonomic slide out tray
  • Included GFP and RFP emission filters to enable the most common fluorescence applications
  • Powerful vision works software allows flexibility of creating custom one-touch workflows or using preinstalled one-touch application templates
  • Generates unmodified raw data. Users have the choice of applying image enhancement tools
For researchers with limited focus on, or just starting in vivo pre-clinical studies, it provides highly sensitive fluorescence in vivo imaging at an affordable price. Camera and light source options provide maximum flexibility in system configuration.
It can support imaging of any probe in the visible to near infrared (NIR) range (400 to 900 nm). NIR imaging minimizes skin autofluorescence (~650 nm) and allows 3x to 8x deeper penetration. The wide spectrum also offers more options for multiplex labeling. This capability enables visualization of tissue/tissue interaction, enhanced signal to background ratio, and detection of multiple fluorescent proteins or labels.
Each camera is configured with it to enable rapid in vivo imaging and repeat observation with minimal disturbance to the subject. The included vision works software tools provide users with the freedom to apply image enhancement and analysis features when needed. They create uncompromised raw data and preserve this true data, promising the highest quantitative value. The enhancement and analysis features offered include many powerful tools, such as: colony counting, area density, noise reduction, background subtraction, inversion, pseudocolor, compositing, and more.
Several components are combined in the optional UVP anesthesia system in order to regulate and administer a combination of oxygen and isoflurane/sevoflurane gas to the animal. The initial anesthesia is performed in the induction chamber. The animal is then moved to the warming plate inside the iBox darkroom and connected to a nose cone. The warming plate generates uniform temperature conditions which maintain a safe body temperature of the animal during the imaging process. A low profile breathing device on the plate connects to patented valves. The system’s non-rebreathing technology prevents backflow of gases into the darkroom and ensures user safety.
Ordering information: Optional UVP anesthesia kit can be ordered for immobilizing small animals.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty