Would you like to log in again?
jetOPTIMUS® DNA transfection Reagent
Supplier: Polyplus-transfection
jetOPTIMUS®
101000051 101000006 101000051SAMPLE 101000051 101000025 101000025 101000006
76299-628EA 0.03 USD InStock
CA76299-630 76299-628 CA76299-632 CA76299-634 76299-634 76299-632 76299-630
jetOPTIMUS® DNA transfection Reagent
Transfection Reagents
jetOPTIMUS® is a best-in-class DNA transfection reagent, based on a novel cationic nanotechnology allowing high efficiency with low volume of reagent. Reach higher transfection efficiency while doubling the number of transfection per vial!
  • Highly efficient: reach maximal gene expression in hard-to-transfect cells
  • Cost-effective: use minimum reagent volume and DNA quantity
  • Biologically relevant: keep an excellent cell viability & morphology
  • Time-saving: transfect with an optimized ready-to-use protocol
jetOPTIMUS® transfection reagent improves cellular uptake and endosomal escape of DNA in cells (even in hard-to-transfect cells) resulting in higher transfection efficiency. In order to work in relevant physiological conditions, transfection with jetOPTIMUS® requires a minimum DNA quantity and reagent volume to keep an excellent cell viability and morphology.
Certifications: Free of charge samples are available upon request. Please contact your local Avantor sales office for further details. Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty