Would you like to log in again?
NEB Cloning Competent E. coli Sampler, New England Biolabs
Supplier: New England Biolabs
C1010S C1010S
76299-644EA 135 USD InStock
CA76299-644 76299-644
NEB Cloning Competent E. coli Sampler, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
The NEB Cloning Competent E.coli Sampler allows to sample four of popular chemically competent strains.
  • NEB 5-alpha: strain for basic cloning; Derivative of the DH5
  • NEB 10-beta: derivative of the DH10B
  • NEB Stable: Ideal for isolation of plasmid clones containing repeat elements and unstable inserts
  • NEB Turbo: Features fast colony growth (6.5 hours) and tight expression control (lacIq)
The Sampler includes two 0.05 ml tubes of each of the following strains (as well as outgrowth medium and control).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty