Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Miller® TurboLite™ EXTREME Sharp Edge Personal Fall Limiters, Honeywell Safety
Supplier: Miller by Honeywell
Miller TurboLite™ edge
MFLEC2-12/6FT MFLEC2-11/9FT MFLEC-3/6FT MFLEC-12/6FT MFLEC-18/9FT MFLEC2-4/6FT MFLEC2-12/9FT MFLEC-11/6FT MFLEC2-11/6FT MFLEC-4/6FT MFLEC2-3/9FT MFLEC-12/9FT MFLEC2-3/6FT MFLEC2-4/9FT MFLEC-11/9FT MFLEC2-18/9FT MFLEC-3/9FT MFLEC2-18/6FT MFLEC-4/9FT MFLEC-18/6FT
76299-704EA 483.16 USD InStock
76299-704 76299-694 76299-660 76299-682 76299-692 76299-680 76299-690 76299-658 76299-702 76299-668 76299-678 76299-700 76299-666 76299-676 76299-698 76299-664 76299-674 76299-696 76299-662 76299-684
Miller® TurboLite™ EXTREME Sharp Edge Personal Fall Limiters, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
Cable lifeline for sharp edge applications.
  • Designed for use in sharp edge applications with radius >0.005" (0.13 mm)
  • Galvanized cable lifeline
  • Available in lengths of 6 and 9'
  • Single and twin configurations
  • Variety of lifeline connector options
  • Versatile, reliable, durable, simple, and safe
Miller® TurboLite™ personal fall limiters (PFLs) are designed for performance in edge applications, offering versatility, enhanced user mobility, and maximum safety. These devices maximize versatility for virtually any short-range application and provide protection for a wider range of workers. The quick and easy installation creates a better work experience while increasing safety and lowering the cost of ownership. These devices ensure compatibility with all Miller® harnesses. It is equipped with an easy to install shock-absorbing connector system and galvanized cable lifeline.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty