Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Miller® TurboLite™ T-BAK EXTREME Sharp Edge Personal Fall Limiters, Honeywell Safety
Supplier: Miller by Honeywell
Miller TurboLite™ edge
MFLEC-20/8FT MFLEC2-20/8FT MFLEC-20/11FT MFLEC2-20/11FT
76299-672EA 425.73 USD InStock
76299-672 76299-670 76299-688 76299-686
Miller® TurboLite™ T-BAK EXTREME Sharp Edge Personal Fall Limiters, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
Cable lifeline with web tie-back for sharp edge tie-back applications.
  • Designed for use in sharp edge, tie-back applications with radius ≥0.005" (0.13 mm)
  • Galvanized cable lifeline with Vectran™ web tie-back
  • Available in lengths of 8 and 11'
  • Single and twin configurations
Miller® TurboLite™ Personal Fall Limiters (PFLs) are designed for performance in edge applications, offering versatility, enhanced user mobility, and maximum safety. These devices maximize versatility for virtually any short-range application and provide protection for a wider range of workers. The quick and easy installation creates a better work experience while increasing safety and lowering the cost of ownership. These devices ensure compatibility with all Miller® harnesses. Equipped with an easy to install shock-absorbing connector system and galvanized cable lifeline with a web tie-back extension.
Certifications: Meets OSHA 1926.502, ANSI A10.32, ANSI Z359.14, Class B and LE (Leading Edge) requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty